Anti-SSX2IP Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:40-1:150
Type:
Antigen: SSX2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77710-517
- 77710-518
- Antigen Synonyms:
- MGC75026|ADIP|Afadin DIL domain-interacting protein|ADIP_HUMAN|SSX 2 interacting protein|Afadin and alpha actinin binding protein|OTTHUMP00000011574|SSX2IP|Afadin- and alpha-actinin-binding protein|Synovial sarcoma X breakpoint 2 interacting protein|SSX2-interacting protein|FLJ10848|SSX2 interacting protein|Afadin DIL domain interacting protein|KIAA0923
- MGC75026|ADIP|Afadin DIL domain-interacting protein|ADIP_HUMAN|SSX 2 interacting protein|Afadin and alpha actinin binding protein|OTTHUMP00000011574|SSX2IP|Afadin- and alpha-actinin-binding protein|Synovial sarcoma X breakpoint 2 interacting protein|SSX2-interacting protein|FLJ10848|SSX2 interacting protein|Afadin DIL domain interacting protein|KIAA0923
- Gene ID:
- Q9Y2D8
- Q9Y2D8
- Immunogen:
- Fusion protein of Human SSX2IP
- Fusion protein of Human SSX2IP
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together