Anti-IGFBP3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: IGFBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77710-456
- 77710-455
- Antigen Synonyms:
- IGFBP-3|Binding protein 29|Insulin-like growth factor binding protein 3 precursor|IBP 3|IBP3|IGFBP 3|IGF-binding protein 3|IBP-3|IGF binding protein 3|BP53|Insulin-like growth factor-binding protein 3|BP 53|Acid stable subunit of the 140 K IGF complex|Binding protein 53|Growth hormone dependent binding protein|IBP3_HUMAN|Insulin Like Growth Factor Binding Protein 3|IGFBP3
- IGFBP-3|Binding protein 29|Insulin-like growth factor binding protein 3 precursor|IBP 3|IBP3|IGFBP 3|IGF-binding protein 3|IBP-3|IGF binding protein 3|BP53|Insulin-like growth factor-binding protein 3|BP 53|Acid stable subunit of the 140 K IGF complex|Binding protein 53|Growth hormone dependent binding protein|IBP3_HUMAN|Insulin Like Growth Factor Binding Protein 3|IGFBP3
- Gene ID:
- P17936
- P17936
- Immunogen:
- Synthetic peptide of Human IGFBP3
- Synthetic peptide of Human IGFBP3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together