Anti-CTSF Rabbit Polyclonal Antibody
New Product
Type:
Antigen: CTSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77710-415
- 77710-416
- Antigen Synonyms:
- CLN13|EC 3.4.22.41|CATSF|Ctsf|CATF_HUMAN|Cathepsin F|AI481912|CathepsinF
- CLN13|EC 3.4.22.41|CATSF|Ctsf|CATF_HUMAN|Cathepsin F|AI481912|CathepsinF
- Gene ID:
- Q9UBX1
- Q9UBX1
- Immunogen:
- Recombinant Human Cathepsin F protein (273-484AA)
- Recombinant Human Cathepsin F protein (273-484AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together