Anti-FES Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: FES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77710-294
- 77710-293
- Antigen Synonyms:
- proto-oncogene tyrosine-protein kinase Fes/Fps|Oncogene FES feline sarcoma virus|cFES|c-fes/fps protein|FES proto oncogene, tyrosine kinase|Proto-oncogene c-Fps|Feline sarcoma (Snyder Theilen) viral (v fes)/Fujinami avian sarcoma (PRCII) viral (v fps) oncogene homolog|Proto oncogene tyrosine protein kinase Fes/Fps|Oncogene FES|feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami|FES_HUMAN|Proto-oncogene c-Fes|fes|Tyrosine-protein kinase Fes/Fps|FPS|P93c fes|Feline sarcoma/Fujinami avian sarcoma oncogene homolog|Feline sarcoma oncogene
- proto-oncogene tyrosine-protein kinase Fes/Fps|Oncogene FES feline sarcoma virus|cFES|c-fes/fps protein|FES proto oncogene, tyrosine kinase|Proto-oncogene c-Fps|Feline sarcoma (Snyder Theilen) viral (v fes)/Fujinami avian sarcoma (PRCII) viral (v fps) oncogene homolog|Proto oncogene tyrosine protein kinase Fes/Fps|Oncogene FES|feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami|FES_HUMAN|Proto-oncogene c-Fes|fes|Tyrosine-protein kinase Fes/Fps|FPS|P93c fes|Feline sarcoma/Fujinami avian sarcoma oncogene homolog|Feline sarcoma oncogene
- Gene ID:
- P07332
- P07332
- Immunogen:
- Fusion protein of Human FES
- Fusion protein of Human FES
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together