Anti-Egln3 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Egln3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77710-100
- 77710-101
- Antigen Synonyms:
- EC 1.14.11.29|HIF-PH3|HIF-prolyl hydroxylase 3|Prolyl hydroxylase domain-containing protein 3|PHD3|Hypoxia-inducible factor prolyl hydroxylase 3|Egln3Egl nine homolog 3|HPH-3|SM-20
- EC 1.14.11.29|HIF-PH3|HIF-prolyl hydroxylase 3|Prolyl hydroxylase domain-containing protein 3|PHD3|Hypoxia-inducible factor prolyl hydroxylase 3|Egln3Egl nine homolog 3|HPH-3|SM-20
- Gene ID:
- Q91UZ4
- Q91UZ4
- Immunogen:
- Recombinant Mouse Egl nine homolog 3 protein (2-239AA)
- Recombinant Mouse Egl nine homolog 3 protein (2-239AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together