Anti-PLXNA1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLXNA1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77709-744
- 77709-745
- Antigen Synonyms:
- NOV|Semaphorin receptor NOV|Plexin-A1|PLXN1|PLXNA1
- NOV|Semaphorin receptor NOV|Plexin-A1|PLXN1|PLXNA1
- Gene ID:
- Q9UIW2
- Q9UIW2
- Immunogen:
- Recombinant Human Plexin-A1 protein (32-296AA)
- Recombinant Human Plexin-A1 protein (32-296AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together