Anti-CCAR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: CCAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77709-486
- 77709-487
- Antigen Synonyms:
- cell division cycle and apoptosis regulator 1|MGC7730|Death inducer with SAP domain|FLJ20734|CCAR1_HUMAN|Cell cycle and apoptosis regulatory protein 1|Ccar1|CARP-1|FLJ10839|FLJ10590|FLJ13376|Cell division cycle and apoptosis regulator protein 1|Carp1|RP11-437A18.1
- cell division cycle and apoptosis regulator 1|MGC7730|Death inducer with SAP domain|FLJ20734|CCAR1_HUMAN|Cell cycle and apoptosis regulatory protein 1|Ccar1|CARP-1|FLJ10839|FLJ10590|FLJ13376|Cell division cycle and apoptosis regulator protein 1|Carp1|RP11-437A18.1
- Gene ID:
- Q8IX12
- Q8IX12
- Immunogen:
- Fusion protein of Human CCAR1
- Fusion protein of Human CCAR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together