Anti-PHKB Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: PHKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77709-449
- 77709-448
- Antigen Synonyms:
- Phosphorylase kinase subunit beta|DKFZp781E15103|KPBB_HUMAN|Phosphorylase kinase beta subunit|Phosphorylase kinase beta polypeptide|FLJ41698|Phosphorylase b kinase regulatory subunit beta|PHKB|Phosphorylase kinase B
- Phosphorylase kinase subunit beta|DKFZp781E15103|KPBB_HUMAN|Phosphorylase kinase beta subunit|Phosphorylase kinase beta polypeptide|FLJ41698|Phosphorylase b kinase regulatory subunit beta|PHKB|Phosphorylase kinase B
- Gene ID:
- Q93100
- Q93100
- Immunogen:
- Fusion protein of Human PHKB
- Fusion protein of Human PHKB
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together