Anti-ZSCAN21 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: ZSCAN21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77709-429
- 77709-430
- Antigen Synonyms:
- Zinc finger and SCAN domain containing protein 21|Renal carcinoma antigen NY REN 21|Zipro1|DKFZp434L134|NYREN21|ZSC21_HUMAN|Zfp-38|ZFP38|zinc finger protein 38 (KOX 25)|DKFZp686H10254|Zinc finger and SCAN domain-containing protein 21|zinc finger protein NY REN 21 antigen|Zinc finger protein 38 homolog|NY REN 21|KOX25|ZSCAN21|Renal carcinoma antigen NY-REN-21|ZNF38|zinc finger and SCAN domain containing 21
- Zinc finger and SCAN domain containing protein 21|Renal carcinoma antigen NY REN 21|Zipro1|DKFZp434L134|NYREN21|ZSC21_HUMAN|Zfp-38|ZFP38|zinc finger protein 38 (KOX 25)|DKFZp686H10254|Zinc finger and SCAN domain-containing protein 21|zinc finger protein NY REN 21 antigen|Zinc finger protein 38 homolog|NY REN 21|KOX25|ZSCAN21|Renal carcinoma antigen NY-REN-21|ZNF38|zinc finger and SCAN domain containing 21
- Gene ID:
- Q9Y5A6
- Q9Y5A6
- Immunogen:
- Recombinant Human Zinc finger and SCAN domain-containing protein 21 protein (1-461AA)
- Recombinant Human Zinc finger and SCAN domain-containing protein 21 protein (1-461AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together