Anti-PSME3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: PSME3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77709-371
- 77709-372
- Antigen Synonyms:
- Proteasome activator 28 gamma|REG GAMMA|11S regulator complex gamma subunit|PSME3|REG-gamma|PA28gamma|Proteasome activator 28 subunit gamma|PSME3_HUMAN|Proteasome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki)|Proteasome (prosome, macropain) activator subunit 3|PA28 gamma|11S regulator complex subunit gamma|Ki, PA28 gamma|PA28g|Proteasome activator complex subunit 3|Ki|Activator of multicatalytic protease subunit 3|Ki antigen|Ki nuclear autoantigen|Proteasome activator subunit 3
- Proteasome activator 28 gamma|REG GAMMA|11S regulator complex gamma subunit|PSME3|REG-gamma|PA28gamma|Proteasome activator 28 subunit gamma|PSME3_HUMAN|Proteasome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki)|Proteasome (prosome, macropain) activator subunit 3|PA28 gamma|11S regulator complex subunit gamma|Ki, PA28 gamma|PA28g|Proteasome activator complex subunit 3|Ki|Activator of multicatalytic protease subunit 3|Ki antigen|Ki nuclear autoantigen|Proteasome activator subunit 3
- Gene ID:
- P61289
- P61289
- Immunogen:
- Recombinant Human Proteasome activator complex subunit 3 protein (2-252AA)
- Recombinant Human Proteasome activator complex subunit 3 protein (2-252AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together