Anti-MAP1S Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP1S
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77709-306
- 77709-307
- Antigen Synonyms:
- MAP1S light chain|C19orf5|BPY2IP1|MAP1S_HUMAN|Microtubule-associated protein 8|MAP1S|FLJ10669|Variable charge Y chromosome 2-interacting protein 1|microtubule-associated protein 1S|MAP-1S|VCY2IP1|BPY2-interacting protein 1|VCY2-interacting protein 1|MAP8|MGC133087|VCY2IP-1
- MAP1S light chain|C19orf5|BPY2IP1|MAP1S_HUMAN|Microtubule-associated protein 8|MAP1S|FLJ10669|Variable charge Y chromosome 2-interacting protein 1|microtubule-associated protein 1S|MAP-1S|VCY2IP1|BPY2-interacting protein 1|VCY2-interacting protein 1|MAP8|MGC133087|VCY2IP-1
- Gene ID:
- Q66K74
- Q66K74
- Immunogen:
- Recombinant Human Microtubule-associated protein 1S protein (794-1053AA)
- Recombinant Human Microtubule-associated protein 1S protein (794-1053AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together