Anti-SLC2A2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC2A2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77709-175
- 77709-176
- Antigen Synonyms:
- GTT2|GLUT2|Glucose transporter type 2|GLUT-2|Solute carrier family 2, facilitated glucose transporter member 2|Glucose Transporter GLUT2|Solute carrier family 2 (facilitated glucose transporter) member 2|SLC2A2|GTR2_HUMAN|Glucose Transporter 2|liver|Glucose transporter, liver/islet|Glucose transporter type 2 liver|Solute carrier family 2 facilitated glucose transporter member 2
- GTT2|GLUT2|Glucose transporter type 2|GLUT-2|Solute carrier family 2, facilitated glucose transporter member 2|Glucose Transporter GLUT2|Solute carrier family 2 (facilitated glucose transporter) member 2|SLC2A2|GTR2_HUMAN|Glucose Transporter 2|liver|Glucose transporter, liver/islet|Glucose transporter type 2 liver|Solute carrier family 2 facilitated glucose transporter member 2
- Gene ID:
- P11168
- P11168
- Immunogen:
- Recombinant Human Solute carrier family 2, facilitated glucose transporter member 2 protein (67-174AA)
- Recombinant Human Solute carrier family 2, facilitated glucose transporter member 2 protein (67-174AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together