Anti-AGR2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: IHC:1:200-1:500
Type:
Antigen: AGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77708-982
- 77708-983
- Antigen Synonyms:
- AG2|secreted cement gland homolog|anterior gradient 2 homolog|HPC 8|HEL S 116|hAG2|AGR 2|hAG-2|hPC8|Epididymis secretory protein Li 116|AG 2|GOB 4|Xenopus|Secreted cement gland protein XAG2 homolog|PDIA17|AG-2|Anterior gradient protein 2 homolog|GOB4|AGR2|Protein disulfide isomerase family A member 17|Anterior gradient 2|HAG2/R|XAG 2|Anterior gradient 2 homolog (Xenopus laevis)|Secreted cement gland protein XAG 2 homolog|AG2 protein|XAG2|AG 2 protein|protein disulphide isomerase family member|Secreted cement gland protein XAG-2 homolog|HAG 2|AGR2_HUMAN|homolog of|Anterior gradient homolog 2
- AG2|secreted cement gland homolog|anterior gradient 2 homolog|HPC 8|HEL S 116|hAG2|AGR 2|hAG-2|hPC8|Epididymis secretory protein Li 116|AG 2|GOB 4|Xenopus|Secreted cement gland protein XAG2 homolog|PDIA17|AG-2|Anterior gradient protein 2 homolog|GOB4|AGR2|Protein disulfide isomerase family A member 17|Anterior gradient 2|HAG2/R|XAG 2|Anterior gradient 2 homolog (Xenopus laevis)|Secreted cement gland protein XAG 2 homolog|AG2 protein|XAG2|AG 2 protein|protein disulphide isomerase family member|Secreted cement gland protein XAG-2 homolog|HAG 2|AGR2_HUMAN|homolog of|Anterior gradient homolog 2
- Gene ID:
- O95994
- O95994
- Immunogen:
- Recombinant Human Anterior gradient protein 2 homolog protein (21-175AA)
- Recombinant Human Anterior gradient protein 2 homolog protein (21-175AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together