Anti-GLI1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77708-931
- 77708-930
- Antigen Synonyms:
- Zfp5|Glioma-associated oncogene|GLI Kruppel family member 1|GLI|Gli 1|GLI family zinc finger 1|Glioma associated oncogene homolog|gli1|Glioma associated oncogene 1|Zinc finger protein GLI1|Glioma associated oncogene homolog 1 (zinc finger protein)|Oncogene GLI|Zfp 5|GLI1_HUMAN|Zinc finger protein GLI 1
- Zfp5|Glioma-associated oncogene|GLI Kruppel family member 1|GLI|Gli 1|GLI family zinc finger 1|Glioma associated oncogene homolog|gli1|Glioma associated oncogene 1|Zinc finger protein GLI1|Glioma associated oncogene homolog 1 (zinc finger protein)|Oncogene GLI|Zfp 5|GLI1_HUMAN|Zinc finger protein GLI 1
- Gene ID:
- P08151
- P08151
- Immunogen:
- Recombinant Human Zinc finger protein GLI1 protein (921-1106AA)
- Recombinant Human Zinc finger protein GLI1 protein (921-1106AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together