Anti-COX6B2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: COX6B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77708-796
- 77708-797
- Antigen Synonyms:
- COX VIb-2|Cytochrome c oxidase subunit VIb isoform 2|Cancer/testis antigen 59|Cytochrome c oxidase subunit VIb|testis-specific isoform|COX6B2Cytochrome c oxidase subunit 6B2|CT59
- COX VIb-2|Cytochrome c oxidase subunit VIb isoform 2|Cancer/testis antigen 59|Cytochrome c oxidase subunit VIb|testis-specific isoform|COX6B2Cytochrome c oxidase subunit 6B2|CT59
- Gene ID:
- Q6YFQ2
- Q6YFQ2
- Immunogen:
- Fusion protein of Human COX6B2
- Fusion protein of Human COX6B2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together