Anti-EPHB1/EPHB2/EPHB3/EPHB4 Rabbit Polyclonal Antibody
77702-201
New Product
- Form:Liquid
- Gene ID:P54762/P29323/P54753/P54760
- Antigen Synonyms:EPHB1|EPH-like kinase 6|HEK6|NET|EPH tyrosine kinase 2|ERK|Neuronally-expressed EPH-related tyrosine kinase|EPHT3|EK6|EPTH3|EPHT2|ELK|Ephrin type-B receptor 1|Tyrosine-protein kinase receptor EPH-2|EPHB2|DRT
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human EphB1/2/3/4 around the non-phosphorylation site of Y600/602/614/596.
- Purification:Antigen affinity purified
- Cat. No.:77702-201
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: EPHB1/EPHB2/EPHB3/EPHB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse