Anti-PIKFYVE Rabbit Polyclonal Antibody
77702-173
New Product
- Form:Liquid
- Gene ID:Q9Y2I7
- Antigen Synonyms:S. cerevisiae|Phosphatidylinositol 3-phosphate 5-kinase type III|Phosphatidylinositol 3 phosphate/phosphatidylinositol 5 kinase type III|PIPkin III|FYVE domain containing 29|PIP5K|FYVE finger containing phosphoinositide kinase|Zinc finger|Phosphatidylinositol 3 phosphate 5 kinase type III|1-phosphatidylinositol 3-phosphate 5-kinase|PtdIns(4)P 5 kinase|Phosphatidylinositol 3-phosphate 5-kinase|ZFYVE29|Phosphoinositide kinase|FYVE finger containing|FYVE finger-containing phosphoinositide kinase|PIPkin-III|CFD|PIP5K3|FAB1|FYV1_HUMAN|1 phosphatidylinositol 3 phosphate 5 kinase|KIAA0981|Type III PIP kinase|homolog of|p235|Phosphatidylinositol 4 phosphate 5 kinase type III|1 phosphatidylinositol 4 phosphate 5 kinase|PIKfyve|epididymis luminal protein 37|MGC40423
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human PIP5KIII around the non-phosphorylation site of S307.
- Purification:Antigen affinity purified
- Cat. No.:77702-173
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: PIKFYVE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat