Anti-CELSR3 Rabbit Polyclonal Antibody
Catalog # 77702-119
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q9NYQ7
- Antigen Synonyms:RESDA1|Multiple epidermal growth factor-like domains protein 2|Multiple EGF-like domains protein 2|Flamingo|Flamingo homolog 1|FMI1|Anchor protein|CELR3_HUMAN|Celsr3|MEGF2|Cadherin family member 11|CDHF11|hFmi1|Epidermal growth factor-like protein 1|Cadherin EGF LAG seven-pass G-type receptor 3|EGFL1|EGF like domain multiple 1|EGF-like protein 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CDHF11.
- Purification:Antigen affinity purified
- Cat. No.:77702-119
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: CELSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human