Anti-S1PR1 (phospho-T236) Rabbit Polyclonal Antibody
77702-109
New Product
- Form:Liquid
- Gene ID:P21453
- Antigen Synonyms:CHEDG 1|CD363|S1PR1_HUMAN|G protein coupled sphingolipid receptor|CHEDG1|Sphingosine 1 phosphate receptor Edg 1|Sphingosine 1-phosphate receptor 1|Endothelial differentiation sphingolipid G protein coupled receptor 1|S1P receptor 1|S1P receptor Edg-1|ECGF 1|Sphingosine 1phosphate receptor type 1 S1P1|S1P receptor Edg 1|S1P(1) receptor|ECGF1|D1S3362|Sphingosine 1 phosphate receptor EDG1|s1pr1|Sphingosine 1-phosphate receptor Edg-1|Endothelial differentiation G-protein coupled receptor 1|g protein-coupled receptor edg-1|FLJ58121|EDG 1|sphingosine 1- phosphate receptor 1|endothelial differentiation G protein coupled receptor 1|EDG1|S1P receptor Edg1|sphingolipid g-protein-coupled receptor 1|S1P1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human EDG-1 around the phosphorylation site of T236.
- Purification:Antigen affinity purified
- Cat. No.:77702-109
- Packaging:Vial
IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: S1PR1 (phospho-T236)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat