Anti-ETHE1 Rabbit Polyclonal Antibody
Catalog # 77687-613
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:O95571
- Antigen Synonyms:Protein ETHE1 mitochondrial|Ethylmalonic encephalopathy protein 1|Ethe1|HSCO|ETHE1 protein|YF13H12|mitochondrial|Protein ETHE1|ethylmalonic encephalopathy 1|mitochondrial precursor|hepatoma subtracted clone one|ETHE1_HUMAN|Hepatoma subtracted clone one protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Persulfide dioxygenase ETHE1, mitochondrial protein (8-254AA)
- Purification:>95%, Protein G purified
- Cat. No.:77687-613
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ETHE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse