Anti-DPYSL2 (phospho-S522) Rabbit Polyclonal Antibody
Catalog # 77702-089
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q16555
- Antigen Synonyms:Dihydropyrimidinase-related protein 2|DHPRP 2|DPYSL 2|ULIP 2 protein|CRMP2|Musunc33|Ulip2|Dpysl2|CRMP-2|Musunc 33|ULIP-2|Dihydropyrimidinase like 2|DPYL 2|TOAD64|Dihydropyrimidinase 2|Collapsin response mediator protein|DPYL2_HUMAN|TOAD 64|CRAM|DRP-2|Unc-33-like phosphoprotein 2|Collapsin response mediator protein hCRMP 2|Collapsin response mediator protein 2|Dihydropyrimidinase related protein 2|Dihydropyrimidinase like 2 long form|DRP2|CRMP 2|N2A3|DHPRP2|DPYL2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human CRMP-2 around the phosphorylation site of S522.
- Purification:Antigen affinity purified
- Cat. No.:77702-089
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: DPYSL2 (phospho-S522)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat