Anti-DOK1 Rabbit Polyclonal Antibody
Catalog # 77702-088
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q99704
- Antigen Synonyms:p62(dok)|Docking protein 1 62kD|DOK1_HUMAN|Downstream of tyrosine kinase 1|P62DOK|DOK 1|pp62|Docking protein 1|DOK1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Dok-1 around the non-phosphorylation site of Y398.
- Purification:Antigen affinity purified
- Cat. No.:77702-088
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: DOK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat