Anti-EPRS Rabbit Polyclonal Antibody
77687-600
New Product
- Form:Liquid
- Gene ID:P07814
- Antigen Synonyms:Bifunctional glutamate/proline tRNA ligase|Prolyl tRNA synthetase|PARS|Glutamate tRNA ligase|Cell proliferation-inducing gene 32 protein|Glutamyl tRNA synthetase|Bifunctional aminoacyl-tRNA synthetase|PIG 32|Proline--tRNA ligase|QPRS|Eprs|Glu pro tRNA synthetase|DKFZp313B047|QARS|GLNS|Proliferation inducing protein 32|Glutamyl prolyl tRNA synthetase|Glutamatyl prolyl tRNA synthetase|GLUPRORS|Bifunctional aminoacyl tRNA synthetase|Prolyl-tRNA synthetase|Glutamyl-tRNA synthetase|GluRS|Proline tRNA ligase|SYEP_HUMAN|PIG32|EARS|Proliferation inducing gene 32 protein|Glutaminyl tRNA synthetase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Bifunctional glutamate/proline--tRNA ligase protein (1207-1399AA)
- Purification:>95%, Protein G purified
- Cat. No.:77687-600
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:200
Type:
Antigen: EPRS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human