Anti-ENTPD2 Rabbit Polyclonal Antibody
Catalog # 77687-589
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q9Y5L3
- Antigen Synonyms:ecto ATPase 2|Ecto-ATPDase 2|Entpd2|CD39L1|CD39like1|CD39 antigen-like 1|CD39 antigen like 1|ENTP2_HUMAN|ectoATPase 2|CD39 like1|Ecto-ATP diphosphohydrolase 2|ecto ATPDase 2|Ecto-ATPase 2|ecto ATP diphosphohydrolase 2|NTPDase2|Ectonucleoside triphosphate diphosphohydrolase 2|ectoATPDase 2|NTPDase 2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 2 protein (28-268AA)
- Purification:Antigen affinity purified
- Cat. No.:77687-589
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: ENTPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human