Anti-AKR1C2 Rabbit Polyclonal Antibody
Catalog # 77702-048
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P52895
- Antigen Synonyms:AKR1C pseudo|MCDR 2|Chlordecone reductase homolog|3 alpha hydroxysteroid dehydrogenase type III|AK1C2_HUMAN|DD|Type III 3 alpha hydroxysteroid dehydrogenase|BABP|DDH2|SRXY8|Bile acid binding protein|DD2|Pseudo chlordecone reductase|Chlordecone reductase homolog HAKRD|Type II dihydrodiol dehydrogenase|Trans-1|Aldo keto reductase family 1 member C2|3-alpha-HSD3|2-dihydrobenzene-1|2-diol dehydrogenase|DD/BABP|HBAB|3 alpha HSD3|DD-2|Dihydrodiol dehydrogenase/bile acid binding protein|Dihydrodiol dehydrogenase/bile acid-binding protein|MCDR2|AKR1C2|HAKRD|FLJ53800|OTTHUMP00000044759|DDH 2|Aldo-keto reductase family 1 member C2|Type III 3-alpha-hydroxysteroid dehydrogenase|Trans 1 2 dihydrobenzene 1 2 diol dehydrogenase|Dihydrodiol dehydrogenase 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human DD2.
- Purification:Antigen affinity purified
- Cat. No.:77702-048
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: AKR1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human