Anti-ZNF600 Rabbit Polyclonal Antibody
Catalog # 77702-040
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q6ZNG1
- Antigen Synonyms:ZNF600Zinc finger protein 600
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ZNF600.
- Purification:Antigen affinity purified
- Cat. No.:77702-040
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: ZNF600
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human