Anti-EEF1A2 Rabbit Polyclonal Antibody
77687-562
New Product
- Form:Liquid
- Gene ID:Q05639
- Antigen Synonyms:HS1|STN|EF1A|Eef1a2|OTTHUMP00000031776|FLJ41696|STNL|eEF1A-2|EF-1-alpha-2|EEF1AL|Eukaryotic translation elongation factor 1 alpha 2|Elongation factor 1 A 2|EF1A2_HUMAN|Elongation factor 1 alpha|Elongation factor 1 alpha 2|Statin like|EF 1 alpha 2|Statin-S1|Elongation factor 1-alpha 2|Eukaryotic elongation factor 1 A-2|Statin S1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Elongation factor 1-alpha 2 protein (184-463AA)
- Purification:Antigen affinity purified
- Cat. No.:77687-562
- Packaging:Vial
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: EEF1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human