Order Entry
United States
ContactUsLinkComponent
Cloud Point, Diesel (Nominal: -7.7°C), Paragon Scientific, LGC Standards
Cloud Point, Diesel (Nominal: -7.7°C), Paragon Scientific, LGC Standards
Catalog # 77691-401
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Cloud Point, Diesel (Nominal: -7.7°C), Paragon Scientific, LGC Standards
Catalog # 77691-401
Supplier:  LGC Standards
Supplier Number:  ALK-CRMU-CPGO
New Product
CAS Number:  

Specifications

  • Description:
    Cloud Point, Diesel (Nominal: -7.7°C)
  • Size:
    250 mL
  • Cat. No.:
    77691-401

Specifications

About this item