Density, Lubricant (Nominal: 0.86709 g/mL @ 15°C), Paragon Scientific, LGC Standards
Catalog # 77691-405
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Density, Lubricant (Nominal: 0.86709 g/mL @ 15°C)
- Size:250 ml
- Cat. No.:77691-405