$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Viscometry Standards
Type N26, General Purpose Viscosity, Paragon Scientific, LGC Standards
Type N26, General Purpose Viscosity, Paragon Scientific, LGC Standards
Catalog # 77693-378
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Type N26, General Purpose Viscosity, Paragon Scientific, LGC Standards
Catalog # 77693-378
Supplier:
LGC Standards
Supplier Number:
ALK-N26
New Product
CAS Number:
Specifications
Description:
Type N26, General Purpose Viscosity
Size:
500 ml
Cat. No.:
77693-378
Specifications
About this item
Product Details
Product Details