Order Entry
United States
ContactUsLinkComponent
 
Wine - Alcohol (5%), LGC, LGC Standards
  77693-518
New Product
 :  
Wine - Alcohol (5%), LGC, LGC Standards
  77693-518
 :  ERM-BA001
New Product
 :  

 

  • Matrix:
    Wine
  • Description:
    Wine - Alcohol (5%)
  • Size:
    250 mL
  • Cat. No.:
    77693-518