Order Entry
United States
ContactUsLinkComponent
Tool Steel, AISI T1, T12001, 100g, ARMI | MBH, LGC Standards
Tool Steel, AISI T1, T12001, 100g, ARMI | MBH, LGC Standards
Catalog # 77693-605
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Tool Steel, AISI T1, T12001, 100g, ARMI | MBH, LGC Standards
Catalog # 77693-605
Supplier:  LGC Standards
Supplier Number:  IARM-48A-C
New Product
CAS Number:  

Specifications

  • Description:
    Tool Steel, AISI T1, T12001, 100g
  • Size:
    100 g
  • Cat. No.:
    77693-605

Specifications

About this item