Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Kale Powder-Nitrate, LGC, LGC Standards
Kale Powder-Nitrate, LGC, LGC Standards
Kale Powder-Nitrate, LGC, LGC Standards
77696-817
:
LGC Standards
New Product
:
Kale Powder-Nitrate, LGC, LGC Standards
77696-817
:
LGC Standards
:
LGC7114
New Product
:
Matrix:
Kale
Description:
Kale Powder-Nitrate
Size:
10 g
Cat. No.:
77696-817