Order Entry
United States
ContactUsLinkComponent
Ethanol 20mg/100mL in Water, LoGiCal, LGC Standards
Ethanol 20mg/100mL in Water, LoGiCal, LGC Standards
Catalog # 77697-028
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Ethanol 20mg/100mL in Water, LoGiCal, LGC Standards
Catalog # 77697-028
Supplier:  LGC Standards
Supplier Number:  LGCETH-20
New Product
CAS Number:  

Specifications

  • Matrix:
    Water
  • Description:
    Ethanol 20mg/100mL in Water
  • Size:
    1 mL
  • Cat. No.:
    77697-028

Specifications

About this item