$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Petroleum Industry Standards
Reference Gas Oil No. 2 for ASTM D2887, VHG, LGC Standards
Reference Gas Oil No. 2 for ASTM D2887, VHG, LGC Standards
77697-846
:
LGC Standards
New Product
:
Reference Gas Oil No. 2 for ASTM D2887, VHG, LGC Standards
77697-846
:
LGC Standards
:
VHG-RGO-10X1
New Product
:
Matrix:
Reference Gas Oil
Description:
Reference Gas Oil No. 2 for ASTM D2887
Size:
10 x 1 mL
Cat. No.:
77697-846