Reference Gas Oil No. 2 for ASTM D2887, VHG, LGC Standards
Catalog # 77697-847
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Reference Gas Oil
- Description:Reference Gas Oil No. 2 for ASTM D2887
- Size:2fl.oz.
- Cat. No.:77697-847