Viscosity Standard: 120 cSt at 40°C, and 20 cSt at 100°C, VHG, LGC Standards
Catalog # 77695-705
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Viscosity Standard: 120 cSt at 40°C, and 20 cSt at 100°C
- Size:500 ml
- Cat. No.:77695-705