Anti-ABCC11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: ABCC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-863
- 77723-864
- Antigen Symbol:
- ABCC11
- ABCC11
- Antigen Synonyms:
- WW|Multidrug resistance associated protein 8|Multidrug resistance-associated protein 8|ABCC11|ABCCB_HUMAN|ATP binding cassette protein C11|ATP binding cassette transporter sub family C member 11|MRP8|ATP-binding cassette sub-family C member 11|ABCC 11|Multi resistance protein 8|OTTHUMP00000164191|ATP binding cassette sub family C (CFTR/MRP) member 11|ATP binding cassette transporter MRP8|ATP binding cassette sub family C member 11|EWWD
- WW|Multidrug resistance associated protein 8|Multidrug resistance-associated protein 8|ABCC11|ABCCB_HUMAN|ATP binding cassette protein C11|ATP binding cassette transporter sub family C member 11|MRP8|ATP-binding cassette sub-family C member 11|ABCC 11|Multi resistance protein 8|OTTHUMP00000164191|ATP binding cassette sub family C (CFTR/MRP) member 11|ATP binding cassette transporter MRP8|ATP binding cassette sub family C member 11|EWWD
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96J66
- Q96J66
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human ATP-binding cassette sub-family C member 11 protein (1-170AA)
- Recombinant Human ATP-binding cassette sub-family C member 11 protein (1-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together