Anti-PLEKHB2 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLEKHB2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-738
- 77723-739
- Antigen Symbol:
- PLEKHB2
- PLEKHB2
- Antigen Synonyms:
- Plekhb2|Evectin2|FLJ23679|Evectin 2|PH domain-containing family B member 2|Pleckstrin homology domain containing family B member 2|Evectin-2|EVT2|Pleckstrin homology domain-containing family B member 2|FLJ20783|PH domain containing family B member 2|FLJ30436|PKHB2_HUMAN|Pleckstrin homology domain containing, family B (evectins) member 2
- Plekhb2|Evectin2|FLJ23679|Evectin 2|PH domain-containing family B member 2|Pleckstrin homology domain containing family B member 2|Evectin-2|EVT2|Pleckstrin homology domain-containing family B member 2|FLJ20783|PH domain containing family B member 2|FLJ30436|PKHB2_HUMAN|Pleckstrin homology domain containing, family B (evectins) member 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CS7
- Q96CS7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Pleckstrin homology domain-containing family B member 2 protein (1-222AA)
- Recombinant Human Pleckstrin homology domain-containing family B member 2 protein (1-222AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together