Anti-KIAA1191 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200
Type:
Antigen: KIAA1191
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
- Catalog No:
- 77690-356
- 77723-687
- Antigen Symbol:
- KIAA1191
- KIAA1191
- Antigen Synonyms:
- FLJ21022|Kiaa1191|Flavin monooxygenase motif-containing protein of 33 kDa|P60MONOX|Putative monooxygenase p33MONOX|Hypothetical protein LOC57179|Brain-derived rescue factor p60MONOX|P33MX_HUMAN
- FLJ21022|Kiaa1191|Flavin monooxygenase motif-containing protein of 33 kDa|P60MONOX|Putative monooxygenase p33MONOX|Hypothetical protein LOC57179|Brain-derived rescue factor p60MONOX|P33MX_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96A73
- Q96A73
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Putative monooxygenase p33MONOX protein (32-305AA)
- Recombinant Human Putative monooxygenase p33MONOX protein (32-305AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together