Anti-DDX17 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DDX17
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-604
- 77723-603
- Antigen Symbol:
- DDX17
- DDX17
- Antigen Synonyms:
- RH70|DEAD box protein 17|DEAD (Asp Glu Ala Asp) box polypeptide 17 |RNA-dependent helicase p72|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 17|P72|Ddx17|DEAD box protein p72|RNA dependent helicase p72|DEAD box helicase 17|DDX17_HUMAN|Probable ATP-dependent RNA helicase DDX17|DEAD (Asp Glu Ala Asp) box helicase 17
- RH70|DEAD box protein 17|DEAD (Asp Glu Ala Asp) box polypeptide 17 |RNA-dependent helicase p72|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 17|P72|Ddx17|DEAD box protein p72|RNA dependent helicase p72|DEAD box helicase 17|DDX17_HUMAN|Probable ATP-dependent RNA helicase DDX17|DEAD (Asp Glu Ala Asp) box helicase 17
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92841
- Q92841
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable ATP-dependent RNA helicase DDX17 protein (591-692AA)
- Recombinant Human Probable ATP-dependent RNA helicase DDX17 protein (591-692AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together