Anti-NELL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: NELL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Specifications
- Catalog No:
- 77723-597
- 77723-596
- Antigen Symbol:
- NELL1
- NELL1
- Antigen Synonyms:
- FLJ45906|Nel related protein 1|Protein kinase C binding protein NELL1|Protein kinase C-binding protein NELL1|NRP1|NELL 1 protein short isoform|Neural epidermal growth factor like 1|IDH3GL|NEL like protein 1|Nell1|NEL-like protein 1|NEL-like 1 (chicken)|Nel-related protein 1|NEL like 1 (chicken)|NELL1_HUMAN
- FLJ45906|Nel related protein 1|Protein kinase C binding protein NELL1|Protein kinase C-binding protein NELL1|NRP1|NELL 1 protein short isoform|Neural epidermal growth factor like 1|IDH3GL|NEL like protein 1|Nell1|NEL-like protein 1|NEL-like 1 (chicken)|Nel-related protein 1|NEL like 1 (chicken)|NELL1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92832
- Q92832
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein kinase C-binding protein NELL1 protein (160-420AA)
- Recombinant Human Protein kinase C-binding protein NELL1 protein (160-420AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together