Anti-CCT6B Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CCT6B
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-567
- 77723-568
- Antigen Symbol:
- CCT6B
- CCT6B
- Antigen Synonyms:
- CCT-zeta-2|TCP-1-zeta-like|Testis-specific protein TSA303|CCT-zeta-like|T-complex protein 1 subunit zeta-2|CCT6B|TCP-1-zeta-2|Testis-specific Tcp20
- CCT-zeta-2|TCP-1-zeta-like|Testis-specific protein TSA303|CCT-zeta-like|T-complex protein 1 subunit zeta-2|CCT6B|TCP-1-zeta-2|Testis-specific Tcp20
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92526
- Q92526
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human T-complex protein 1 subunit zeta-2 protein (101-330AA)
- Recombinant Human T-complex protein 1 subunit zeta-2 protein (101-330AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together