Anti-MTO1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: MTO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
- Catalog No:
- 77723-547
- 77690-348
- Antigen Symbol:
- MTO1
- MTO1
- Antigen Synonyms:
- MTO1|mitochondrial|CGI-02|Protein MTO1 homolog
- MTO1|mitochondrial|CGI-02|Protein MTO1 homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Z2
- Q9Y2Z2
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein MTO1 homolog, mitochondrial protein (420-680AA)
- Recombinant Human Protein MTO1 homolog, mitochondrial protein (420-680AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together