Anti-ASZ1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ASZ1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-491
- 77723-492
- Antigen Symbol:
- ASZ1
- ASZ1
- Antigen Synonyms:
- SAM and basic leucine zipper domain-containing protein|Germ cell-specific ankyrin|ALP1|Ankyrin-like protein 1|ANKL1|GASZAnkyrin repeat|C7orf7|SAM and basic leucine zipper domain-containing protein 1|ASZ1
- SAM and basic leucine zipper domain-containing protein|Germ cell-specific ankyrin|ALP1|Ankyrin-like protein 1|ANKL1|GASZAnkyrin repeat|C7orf7|SAM and basic leucine zipper domain-containing protein 1|ASZ1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WWH4
- Q8WWH4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1 protein (1-200AA)
- Recombinant Human Ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1 protein (1-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together