Anti-UBE2Q2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2Q2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-464
- 77723-463
- Antigen Symbol:
- UBE2Q2
- UBE2Q2
- Antigen Synonyms:
- UB2Q2_HUMAN|Ubiquitin conjugating enzyme E2Q 2|ubiquitin-conjugating enzyme E2Q (putative) 2|Ubiquitin-conjugating enzyme E2 Q2|Ubiquitin-protein ligase Q2|Ubiquitin conjugating enzyme E2Q family member 2|Ubiquitin carrier protein Q2|UBE2Q2|LOC92912
- UB2Q2_HUMAN|Ubiquitin conjugating enzyme E2Q 2|ubiquitin-conjugating enzyme E2Q (putative) 2|Ubiquitin-conjugating enzyme E2 Q2|Ubiquitin-protein ligase Q2|Ubiquitin conjugating enzyme E2Q family member 2|Ubiquitin carrier protein Q2|UBE2Q2|LOC92912
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVN8
- Q8WVN8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 protein (1-375AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 protein (1-375AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together