Anti-LEO1 Rabbit Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LEO1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-459
- 77723-458
- Antigen Symbol:
- LEO1
- LEO1
- Antigen Synonyms:
- Replicative senescence downregulated leo1 like|LOC123439|RNA polymerase associated protein LEO1|Senescence downregulated leo1 like|LEO1|LEO1_HUMAN|Gm185|Replicative senescence down-regulated leo1-like protein|RNA polymerase-associated protein LEO1|Replicative senescence down regulated leo1 like protein|Paf1/RNA polymerase II complex component
- Replicative senescence downregulated leo1 like|LOC123439|RNA polymerase associated protein LEO1|Senescence downregulated leo1 like|LEO1|LEO1_HUMAN|Gm185|Replicative senescence down-regulated leo1-like protein|RNA polymerase-associated protein LEO1|Replicative senescence down regulated leo1 like protein|Paf1/RNA polymerase II complex component
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVC0
- Q8WVC0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human RNA polymerase-associated protein LEO1 protein (2-24AA)
- Peptide sequence from Human RNA polymerase-associated protein LEO1 protein (2-24AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together