Anti-GRHL3 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRHL3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-388
- 77723-389
- Antigen Symbol:
- GRHL3
- GRHL3
- Antigen Synonyms:
- Transcription factor CP2-like 4|Drosophila|GRHL3_HUMAN|grhl3|Transcription factor CP2 like 4|Grainyhead-like protein 3 homolog|Grainyhead like 3|Sister of mammalian grainyhead protein|SOM|Sister of mammalian grainyhead|Grainyhead like protein 3 homolog|TFCP2L4
- Transcription factor CP2-like 4|Drosophila|GRHL3_HUMAN|grhl3|Transcription factor CP2 like 4|Grainyhead-like protein 3 homolog|Grainyhead like 3|Sister of mammalian grainyhead protein|SOM|Sister of mammalian grainyhead|Grainyhead like protein 3 homolog|TFCP2L4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE85
- Q8TE85
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Grainyhead-like protein 3 homolog protein (31-182AA)
- Recombinant Human Grainyhead-like protein 3 homolog protein (31-182AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together