Anti-DYNC2LI1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: DYNC2LI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Specifications
- Catalog No:
- 77690-341
- 77723-367
- Antigen Symbol:
- DYNC2LI1
- DYNC2LI1
- Antigen Synonyms:
- CGI-60|Cytoplasmic dynein 2 light intermediate chain 1|Dynein 2 light intermediate chain|DYNC2LI1|D2LIC|LIC3|DC2L1_HUMAN|Dynein, cytoplasmic 2, light intermediate chain 1
- CGI-60|Cytoplasmic dynein 2 light intermediate chain 1|Dynein 2 light intermediate chain|DYNC2LI1|D2LIC|LIC3|DC2L1_HUMAN|Dynein, cytoplasmic 2, light intermediate chain 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TCX1
- Q8TCX1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cytoplasmic dynein 2 light intermediate chain 1 protein (1-352AA)
- Recombinant Human Cytoplasmic dynein 2 light intermediate chain 1 protein (1-352AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together