Anti-GABPB2 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: GABPB2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77723-317
- 77723-316
- Antigen Symbol:
- GABPB2
- GABPB2
- Antigen Synonyms:
- Nuclear respiratory factor 2|GABPB 1|GA binding protein beta chain|GABP2_HUMAN|GABP subunit beta-2|BABPB2|GA binding protein transcription factor beta subunit 2|Gabpb2|E4TF1|RP11-68I18.1|E4TF1B|E4TF1 47|Transcription factor E4TF1 53|GABPB-2|Transcription factor E4TF1 47|NRF2B2|GABP subunit beta 1|Nuclear respiratory factor 2 subunit beta|GABPB1|E4TF1 53|GABP subunit beta 2|GA-binding protein subunit beta-2|NRF2B1|GABP 2
- Nuclear respiratory factor 2|GABPB 1|GA binding protein beta chain|GABP2_HUMAN|GABP subunit beta-2|BABPB2|GA binding protein transcription factor beta subunit 2|Gabpb2|E4TF1|RP11-68I18.1|E4TF1B|E4TF1 47|Transcription factor E4TF1 53|GABPB-2|Transcription factor E4TF1 47|NRF2B2|GABP subunit beta 1|Nuclear respiratory factor 2 subunit beta|GABPB1|E4TF1 53|GABP subunit beta 2|GA-binding protein subunit beta-2|NRF2B1|GABP 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TAK5
- Q8TAK5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human GA-binding protein subunit beta-2 protein (163-440AA)
- Recombinant Human GA-binding protein subunit beta-2 protein (163-440AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together